Supreno® EC Powder-Free Nitrile Gloves, Microflex®, Ansell
Supplier: Microflex
SEC-375-S SEC-375-M SEC-375-L SEC-375-XL SEC-375-XXL SEC-375-XS SEC-375-3XL SEC-375-XS SEC-375-XS
CA32916-670CS 323.94 CAD
CA32916-670 CA32916-672 CA32916-674 CA32916-676 CA32916-678 CA10755-078 CA10755-082 76311-656 CA10755-079
Supreno® EC Powder-Free Nitrile Gloves, Microflex®, Ansell
Gloves
Soft, durable nitrile formulation delivers improved comfort, strength and durability for maximum protection against rips, snags or tears
  • Ambidextrous
  • Powder-free
  • Beaded cuffs
  • Online chlorination
Gloves are suitable for high-risk applications and feature an extended cuff to protect the wrist and forearm. Interior polymer coating makes gloves easy to don. Textured fingers allow for reliable grip in wet and dry conditions.
Designed for workers in high risk environments, this glove features a low 0.65 AQL for advanced barrier protection.
Certifications: Gloves meet or exceed the emergency medical glove requirements of NFPA 1999. Tested for use with chemotherapy/cytotoxic drugs.
Packaging: Gloves are packaged 50 gloves per dispenser box.
Caution: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.