Indium ≥99.998% (metals basis), wire, Puratronic®, Ø 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

11462-G1 11462-G5 11462-G9
CAAA11462-G1EA 213.3 CAD
CAAA11462-G1 CAAA11462-G5 CAAA11462-G9
Indium ≥99.998% (metals basis), wire, Puratronic®, Ø 1.0 mm (0.04 in)
Indium

MDL: MFCD00134048


Purity: 99.998+% CAS No.: 7440-74-6 Physical Form: ≈5.74g/m Odor: Odorless MDL No.: MFCD00134048

Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now

Specification Test Results

Assay (by Metals Basis) 99.998+%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR