(±)-1,2-Hexanediol ≥96.0%
Supplier: TCI America
Synonyms:
Hexane-1,2-diol, DL-1,2-Hexanediol, (±)-1,2-Dihydroxy hexane
H0688-25ML
H0688-500ML
H0688-100ML
TCH0688-025MLEA
60.04
CAD
TCH0688-025ML
TCH0688-500ML
TCH0688-100ML
(±)-1,2-Hexanediol ≥96.0%
(±)-1,2-Hexanediol
CAS Number: 6920-22-5
MDL Number: MFCD00010737
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 122
Specific Gravity (20/20): 0.95
Other Product Names / Synonyms: 1,2-Dihydroxyhexane, 1,2-Hexylene Glycol
Formula:
C₆H₁₄O₂ Boiling Pt: 118 °C Melting Pt: 45 °C Density: 0.95 (20 °C) Flash Pt: 122 °C |
MDL Number:
MFCD00010737 CAS Number: 6920-22-5 EINECS: 230-029-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...