α-n-Hexylcinnamaldehyde ≥90.0% (by GC)

Supplier: TCI America
Warning

H0685-500ML H0685-25ML
TCH0685-500MLEA 151.05 CAD
TCH0685-500ML TCH0685-025ML
α-n-Hexylcinnamaldehyde ≥90.0% (by GC)
α-n-Hexylcinnamaldehyde

CAS Number: 101-86-0
MDL Number: MFCD00006989
Molecular Formula: C15H20O
Molecular Weight: 216.32
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Specific Gravity (20/20): 0.96


Product Categories: Bioscience, Other Pharmaceutical Ingredients, Phenylpropanoids
Other Product Names / Synonyms: 2-Hexyl-3-phenylacrylaldehyde, 2-Hexyl-3-phenyl-2-propenal

Formula: C15H20O
Boiling Pt: 175 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00006989
CAS Number: 101-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38c0d51e12203a9f67052057801b2d3d0182d817&vid=38c0d51e12203a9f67052057801b2d3d0182d817&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR