4-Hydroxy-2-quinolinecarboxylic acid hydrate ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxyquinaldic acid hydrate, Kynurenic acid hydrate

H0303-1G H0303-5G
TCH0303-001GEA 85.32 CAD
TCH0303-001G TCH0303-005G
4-Hydroxy-2-quinolinecarboxylic acid hydrate ≥98.0%
4-Hydroxy-2-quinolinecarboxylic acid hydrate

CAS Number: 492-27-3
MDL Number: MFCD00006753
Molecular Formula: C10H7NO3
Molecular Weight: 189.17
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal


Other Product Names / Synonyms: 4-Hydroxyquinaldic Acid, 4-Hydroxyquinoline-2-carboxylic Acid

Formula: C₁₀H₇NO₃·xH₂O
MW: 189.17 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00006753
CAS Number: 345909-35-5
EINECS: 207-751-5
Merck Index: 13,05342
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad26561cc43fdacae6e1e792098b7f4994f72b0d&vid=ad26561cc43fdacae6e1e792098b7f4994f72b0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR