Hydantoin ≥99.0% (by titrimetric analysis)
Supplier: TCI America
H0167-25G
H0167-500G
TCH0167-025GEA
37.92
CAD
TCH0167-025G
TCH0167-500G
Hydantoin ≥99.0% (by titrimetric analysis)
Hydantoin
CAS Number: 461-72-3
MDL Number: MFCD00005259
Molecular Formula: C3H4N2O2
Molecular Weight: 100.08
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 224
Other Product Names / Synonyms: Glycolylurea, 2,4-Imidazolidinedione
Formula:
C₃H₄N₂O₂ Melting Pt: 224 °C |
MDL Number:
MFCD00005259 CAS Number: 461-72-3 EINECS: 207-313-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...