Hydrochloric acid 0.100 N (N/10) in 2-propanol
Supplier: Ricca Chemical
|
Danger
|
R3800000-4A
R3800000-1A
R3800000-20F
R3800000-500A
RC38001EA
534.04
CAD
RC38001
RC380032
RCR3800000-20F
RCR3800000500A
Hydrochloric acid 0.100 N (N/10) in 2-propanol
Hydrochloric acid
Formula:
HCl Density: 0.79 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1219 ADR: 3,II |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sodium Hydroxide/Phenolphthalein) | 0.0995-0.1005 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...