Hydrochloric acid 5.00 N analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

R3740000-4A R3740000-1A R3740000-20F R3740000-10F R3740000-500A
RC37401EA 276.5 CAD
RC37401 RC374032 RC3740-5 RCR3740000-10F RCR3740000500A
Hydrochloric acid 5.00 N analytical reagent
Hydrochloric acid
Formula: HCl
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to slightly greenish-yellow liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 4.995-5.005 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR