Hydrochloric acid 1.00 N

Supplier: Ricca Chemical
Danger

R3700000-10F R3700000-20F R3700000-4A R3700000-1A R3700000-120A R3700000-500A R3700000-120A
RC370025EA 325.48 CAD
RC370025 RC37005 RC37001 RC370032 RC3700-4 CARC3700-16 CARC3700-4
Hydrochloric acid 1.00 N
Hydrochloric acid

Standardized at 25 °C against chemicals whose certification is traceable to NIST.

Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Tris(hydroxymethyl)aminomethane/Methyl Red) 0.999-1.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR