Nitrile Gloves, Honeywell Safety
Supplier: Honeywell Safety
Nitty-Gritty®
LA102G/9 LA102G/11XXL LA172G/9 LA102G/11 LA132G/8M LA132G/11 LA172G/10 LA132G/10 LA132G/9 LA132G/8 LA102G/10XL LA102G/9L LA172G/8 LA132G/7S LA102G/7 LA102G/10 LA172G/7S LA172G/8M LA132G/11XXL LA102G/7S LA102G/8 LA132G/9L LA102G/8M
CA32888-240CS 334.94 CAD
32888-274 CA32888-240 32888-284 32888-240 CA32888-274 32888-272 CA32888-242 32888-282 32888-278 CA32888-280 32888-244 CA32888-270 32888-276 CA32888-282 CA32888-272 32888-286 32888-242 CA32888-248 32888-248 32888-246 CA32888-244 CA32888-278 CA32888-246
Nitrile Gloves, Honeywell Safety
Gloves
Anatomically shaped, flexible gloves feature a straight cuff edge for good tear resistance.
  • Styles with flocked interior give the glove a soft, pleasant feel and absorb perspiration
  • Case hardened for easy donning and added toughness
High nitrile buna rubber (NBR) content provides superior resistance to cuts, snags, abrasion, and punctures. Available in many thicknesses to meet your needs.
Certifications: Comply with USDA and FDA regulations, 21CFR, for use in food processing.