KIMAX® Cylindrical Weighing Bottles, Regular and Tall Form, Kimble®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
15145-1550 15145-2540 15145-2550 15145-3060 15145-4050 15145-4080 15145-5060
16711-020CS 1160.19 CAD
16711-020 16711-063 16711-085 16711-100 16711-121 16711-143 16711-167
KIMAX® Cylindrical Weighing Bottles, Regular and Tall Form, Kimble®, DWK Life Sciences
Bottles Weighing Bottles
Bottles are supplied with a plug-style, closed bottom airtight glass [ST] stopper that fits into an internally ground body.
Bottles also feature a frosted marking area. Designed from Federal Specification NNN-B-1493, Type I, Style I requirements.