Hydrochloric acid 6.0 N, StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Hydrochloric acid solution, Muriatic acid
Management of Change (MOC) category = NI
H168-05
CAMKH16807
MKH16805CS
567.17
CAD
MKH16805
CAMKH16807
Hydrochloric acid 6.0 N, StandARd® volumetric solution, Macron Fine Chemicals™
Hydrochloric acid
Formula:
HCl Density: 1.06 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Normality | 5.95 - 6.05 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...