Hydrochloric acid 1.0 N, StandARd® volumetric solution, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = NI

6388-60 CAMK638801 CAMK638805
MK638860CS 491.89 CAD
MK638860 CAMK638801 CAMK638805
Hydrochloric acid 1.0 N, StandARd® volumetric solution, Macron Fine Chemicals™
Hydrochloric acid

J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications. 


J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications. 

Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.995 - 1.005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR