Hydrochloric acid 0.1 N (N/10), StandARd® Volumetric, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

M.O.C. Category NI

MACR2853-01 MACR2853-05 MACR2853-60
CAMK285301EA 192.18 CAD
CAMK285301 CAMK285305 CAMK285360
Hydrochloric acid 0.1 N (N/10), StandARd® Volumetric, Macron Fine Chemicals™
Hydrochloric acid
Formula: HCl
MW: 36.46
Density: 1.004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0995 - 0.1005


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR