Hydrochloric acid 36.5 - 38.0% NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Management of Change (MOC) category = R
BAKR9544-08
BAKR9544-02
BAKR9544-33
BAKR9544-05
9544-33
9544-07
9544-05
9544-03
CAJT9544-15EA
6565.47
CAD
CAJT9544-15
CAJT9544-8
CAJT9544-03
CAJT9544-2
CAJT9544-33
CAJT9544-5
JT9544-33
JT9544-7
JT9544-5
JT9544-3
Hydrochloric acid 36.5 - 38.0% NF, Multi-Compendial, J.T.Baker®
Hydrochloric acid
Corrosive liquid.
Formula:
HCl Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Meets J.P. Chemical Specifications | |
Meets B.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
Meets N.F. Requirements | |
GMP Manufactured Product | |
Store below 25°C | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
NF - Identification | Passes Test |
NF - Residue on Ignition | ≤ 0.008 % |
NF - Bromide or Iodide | Passes Test |
NF - Free Bromine or Chlorine | Passes Test |
NF - Sulfate (SO₄) | Passes Test |
NF - Sulfite (SO₃) | Passes Test |
NF - Assay (HCl) | 36.5 - 38.0 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Identification C | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Free Chlorine (as Cl₂) | ≤ 4 ppm |
EP/BP - Sulfate (SO₄) | ≤ 20 ppm |
EP/BP - Residue on Evaporation | ≤ 0.01 % |
EP/BP - Assay (HCl) | 35.0 - 39.0 % |
JP - Assay (HCl) | 35.0 - 38.0 % |
JP - Identification | Passes Test |
JP - Residue on ignition, mg/10mL | ≤ 1.0 |
JP - Bromide or Iodide | Passes Test |
JP - Bromine or Chlorine | Passes Test |
JP - Sulfate (SO₄) | Passes Test |
JP - Sulfite (SO₃) | Passes Test |
No Class 1, 2, 3 or other solvents are used or produced in the manufacturing or purification of the product. | ≤ 1 ppm |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...