Hydrochloric acid 37,0 - 38,0%, CMOS Microelectronic, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

M.O.C. Category R

9539-03 BAKR9539-03 BAKR9539-33 BAKR9539-05
JT9539-3CS 1142.74 CAD
JT9539-3 CAJT9539-3 CAJT9539-33 CAJT9539-5 CAJT9539-01
Hydrochloric acid 37,0 - 38,0%, CMOS Microelectronic, J.T.Baker®
Hydrochloric acid
Formula: HCl
MW: 36.46
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

For Microelectronic Use
Store below 25°C
Assay (as HCl) (by acid-base titrn) 37.0 - 38.0 %
Color (APHA) ≤ 10
Residue after Ignition ≤ 3 ppm
Extractable Organic Substances ≤ 3 ppm
Bromide (Br) ≤ 0.005 %
Free Halogen (as Cl₂) Passes Test
Ammonium (NH₄) ≤ 1 ppm
Phosphate (PO₄) ≤ 0.05 ppm
Sulfate (SO₄) ≤ 0.3 ppm
Sulfite (SO₃) ≤ 0.8 ppm
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Arsenic and Antimony (as As) ≤ 5.0 ppb
Trace Impurities - Barium (Ba) ≤ 20.0 ppb
Trace Impurities - Beryllium (Be) ≤ 10.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 20.0 ppb
Trace Impurities - Boron (B) ≤ 20.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 5.0 ppb
Trace Impurities - Calcium (Ca) ≤ 100.0 ppb
Trace Impurities - Chromium (Cr) ≤ 10.0 ppb
Trace Impurities - Cobalt (Co) ≤ 5.0 ppb
Trace Impurities - Copper (Cu) ≤ 5.0 ppb
Trace Impurities - Gallium (Ga) ≤ 20.0 ppb
Trace Impurities - Germanium (Ge) ≤ 20 ppb
Trace Impurities - Gold (Au) ≤ 20 ppb
Heavy Metals (as Pb) ≤ 100 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Lead (Pb) ≤ 20.0 ppb
Trace Impurities - Lithium (Li) ≤ 20.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Manganese (Mn) ≤ 5.0 ppb
Trace Impurities - Molybdenum (Mo) ≤ 10.0 ppb
Trace Impurities - Nickel (Ni) ≤ 10.0 ppb
Trace Impurities - Niobium (Nb) ≤ 10.0 ppb
Trace Impurities - Potassium (K) ≤ 50 ppb
Trace Impurities - Silicon (Si) ≤ 100.0 ppb
Trace Impurities - Silver (Ag) ≤ 20.0 ppb
Trace Impurities - Sodium (Na) ≤ 100.0 ppb
Trace Impurities - Strontium (Sr) ≤ 20.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 10.0 ppb
Trace Impurities - Thallium (Tl) ≤ 20.0 ppb
Trace Impurities - Tin (Sn) ≤ 50 ppb
Trace Impurities - Titanium (Ti) ≤ 10.0 ppb
Trace Impurities - Vanadium (V) ≤ 10.0 ppb
Trace Impurities - Zinc (Zn) ≤ 30.0 ppb
Trace Impurities - Zirconium (Zr) ≤ 10.0 ppb
Particle Count - 0.5 µm and greater ≤ 50 par/ml
Particle Count - 1.0 µm and greater ≤ 10 par/ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR