Synthware Erlenmeyer Flasks, Kemtech America
Supplier: Kemtech America
F661405 F661410 F661425 F661450 F661950 F662450 F662950 F661125 F669125 F664125 F664250 F669250 F664500 F669500 F664000 F668000
60003-012EA 36.13 CAD
60003-012 60003-014 60003-018 60003-022 60003-024 60003-026 60003-028 60003-030 60003-032 60003-034 60003-038 60003-040 60003-042 60003-044 60003-046 60003-048
Synthware Erlenmeyer Flasks, Kemtech America
Flasks Erlenmeyer Flasks
With [ST] joint.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Various sizes available
All sizes are hand-blown from tubing to ensure uniform, heavy wall. Stopper is not supplied.