Hydrochloric acid 36,5 - 38,0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

BAKR9530-00 BAKR9530-33
CAJT9530-0CS 983.46 CAD
CAJT9530-0 CAJT9530-33
Hydrochloric acid 36,5 - 38,0%, BAKER INSTRA-ANALYZED® for trace metal analysis, J.T.Baker®
Hydrochloric acid

For trace metal analysis. Certificate of lot analysis provided.

Formula: HCl
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Store below 25°C
ACS - Assay (as HCl) (by acid-base titrn) 36.5 - 38.0 %
ACS - Color (APHA) ≤ 10
ACS - Residue after Ignition ≤ 3 ppm
ACS - Specific Gravity at 60°/60°F 1.185 - 1.192
ACS - Bromide (Br) ≤ 0.005 %
ACS - Extractable Organic Substances ≤ 5 ppm
ACS - Free Chlorine (as Cl₂) ≤ 0.5 ppm
Phosphate (PO₄) ≤ 0.05 ppm
Sulfate (SO₄) ≤ 0.5 ppm
Sulfite (SO₃) ≤ 0.8 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Arsenic (As) ≤ 0.010 ppm
Trace Impurities - Aluminum (Al) ≤ 10.0 ppb
Arsenic and Antimony (as As) ≤ 5.0 ppb
Trace Impurities - Barium (Ba) ≤ 1.0 ppb
Trace Impurities - Beryllium (Be) ≤ 1.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 10.0 ppb
Trace Impurities - Boron (B) ≤ 20.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 1.0 ppb
Trace Impurities - Calcium (Ca) ≤ 50.0 ppb
Trace Impurities - Chromium (Cr) ≤ 1.0 ppb
Trace Impurities - Cobalt (Co) ≤ 1.0 ppb
Trace Impurities - Copper (Cu) ≤ 1.0 ppb
Trace Impurities - Gallium (Ga) ≤ 1.0 ppb
Trace Impurities - Germanium (Ge) ≤ 3.0 ppb
Trace Impurities - Gold (Au) ≤ 4.0 ppb
Heavy Metals (as Pb) ≤ 100 ppb
Trace Impurities - Iron (Fe) ≤ 15 ppb
Trace Impurities - Lead (Pb) ≤ 1.0 ppb
Trace Impurities - Lithium (Li) ≤ 1.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 10.0 ppb
Trace Impurities - Manganese (Mn) ≤ 1.0 ppb
Trace Impurities - Mercury (Hg) ≤ 0.5 ppb
Trace Impurities - Molybdenum (Mo) ≤ 10.0 ppb
Trace Impurities - Nickel (Ni) ≤ 4.0 ppb
Trace Impurities - Niobium (Nb) ≤ 1.0 ppb
Trace Impurities - Potassium (K) ≤ 9.0 ppb
Trace Impurities - Selenium (Se), For Information Only
Trace Impurities - Silicon (Si) ≤ 100.0 ppb
Trace Impurities - Silver (Ag) ≤ 1.0 ppb
Trace Impurities - Sodium (Na) ≤ 100.0 ppb
Trace Impurities - Strontium (Sr) ≤ 1.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 1.0 ppb
Trace Impurities - Thallium (Tl) ≤ 5.0 ppb
Trace Impurities - Tin (Sn) ≤ 5.0 ppb
Trace Impurities - Titanium (Ti) ≤ 1.0 ppb
Trace Impurities - Vanadium (V) ≤ 1.0 ppb
Trace Impurities - Zinc (Zn) ≤ 5.0 ppb
Trace Impurities - Zirconium (Zr) ≤ 1.0 ppb
Product Information (not specifications):
Appearance (clear, fuming liquid)
Meets ACS Specifications


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR