PYREX® VISTA™ Erlenmeyer Flasks, Narrow Mouth, Corning
Supplier: Corning
70980-1L 70980-125 70980-250 70980-25 70980-500 70980-50
89090-302CS 384.26 CAD
89090-302 89090-316 89090-348 89090-360 89090-386 89090-398
PYREX® VISTA™ Erlenmeyer Flasks, Narrow Mouth, Corning
Flasks Erlenmeyer Flasks
PYREX®, narrow neck.
  • Uniform wall thickness
  • Resistance to thermal shock
  • Approximate graduations
The uniform wall thickness to provide the proper balance between mechanical strength and thermal shock resistance.
these reusable flasks provides an extra-large blue marking space for easy identification. With Beaded top rims.
Certifications: ASTM E-1404 Type 1