n-Hexane ≥95.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = NI

BAKR9304-02 BAKR9304-03 BAKR9304-33
CAJT9304-2CS 1626.06 CAD
CAJT9304-2 CAJT9304-3 CAJT9304-33 CAJT9304-88
n-Hexane ≥95.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
n-Hexane

For use in liquid chromatography and spectrophotometry. UV cutoff 192 nm. Lot analysis on label.

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94.3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.008
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.08
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.2
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤192
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (Total Saturated C₆ Isomers) (by GC, corrected for water) ≥99.5 %
Assay (as n-Hexane) (by GC, corrected for water) ≥95.0 %
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.01 %
Filtered through a 0.2 micron filter.
Packaged under Nitrogen.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR