TE Buffer, pH 8.0, Molecular Biology grade, Calbiochem®

Supplier: MilliporeSigma

8890-100ML
CAEM8890EA 732.94 CAD
CAEM8890
TE Buffer, pH 8.0, Molecular Biology grade, Calbiochem®
Buffers Biological Buffers

Contains 10mM TRIS and 1mM EDTA


  • Conductivity at 25°C: 775 - 975 µmho
  • pH of solution at 25°C: 7.90 - 8.10


DNase-, RNase-, and protease-free.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR