n-Heptane ≥99% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: high purity solvents

Management of Change (MOC) category = N

BAKR9177-33 9177-03
CAJT9177-3EA 2907.9 CAD
CAJT9177-3 CAJT9177-33 JT9177-3
n-Heptane ≥99% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
n-Heptane

For liquid chromatography and spectrophotometry. UV cutoff 197nm. Packaged under nitrogen. Lot analysis on label.

Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4
Melting Pt: -90.6
Density: 0.6838 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤197
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.2 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (by GC, corrected for water) ≥99.0 %
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR