Hydrofluoric acid 47 - 51%, ULTREX® II, Ultrapure, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC


Management of Change (MOC) category = N

BAKR6904-05 6904-05
CAJT6904-5EA 3487.96 CAD
JT6904-5 CAJT6904-5
Hydrofluoric acid 47 - 51%, ULTREX® II, Ultrapure, J.T.Baker®
Hydrofluoric acid
Formula: HF
MW: 20.01 g/mol
MDL Number: MFCD00011346
CAS Number: 7664-39-3
UN: 1790
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (HF)(w/w) 47 - 51 %
Total Sulfur ≤100 ppb
Trace Impurities - Aluminum (Al) ≤20 ppt
Trace Impurities - Antimony (Sb) ≤20 ppt
Trace Impurities - Arsenic (As) ≤50 ppt
Trace Impurities - Barium (Ba) ≤10 ppt
Trace Impurities - Beryllium (Be) ≤10 ppt
Trace Impurities - Bismuth (Bi) ≤10 ppt
Trace Impurities - Boron (B) ≤100 ppt
Trace Impurities - Cadmium (Cd) ≤10 ppt
Trace Impurities - Calcium (Ca) ≤10 ppt
Trace Impurities - Cerium (Ce) ≤10 ppt
Trace Impurities - Cesium (Cs) ≤10 ppt
Trace Impurities - Chromium (Cr) ≤10 ppt
Trace Impurities - Cobalt (Co) ≤10 ppt
Trace Impurities - Copper (Cu) ≤10 ppt
Trace Impurities - Dysprosium (Dy) ≤1 ppt
Trace Impurities - Erbium (Er) ≤1 ppt
Trace Impurities - Europium (Eu) ≤1 ppt
Trace Impurities - Gadolinium (Gd) ≤1 ppt
Trace Impurities - Gallium (Ga) ≤10 ppt
Trace Impurities - Germanium (Ge) ≤10 ppt
Trace Impurities - Gold (Au) ≤20 ppt
Trace Impurities - Hafnium (Hf) ≤10 ppt
Trace Impurities - Holmium (Ho) ≤1 ppt
Trace Impurities - Indium (In) ≤1 ppt
Trace Impurities - Iron (Fe) ≤10 ppt
Trace Impurities - Lanthanum (La) ≤10 ppt
Trace Impurities - Lead (Pb) ≤10 ppt
Trace Impurities - Lithium (Li) ≤10 ppt
Trace Impurities - Lutetium (Lu) ≤1 ppt
Trace Impurities - Magnesium (Mg) ≤10 ppt
Trace Impurities - Manganese (Mn) ≤10 ppt
Trace Impurities - Mercury (Hg) ≤50 ppt
Trace Impurities - Molybdenum (Mo) ≤10 ppt
Trace Impurities - Neodymium (Nd) ≤1 ppt
Trace Impurities - Nickel (Ni) ≤20 ppt
Trace Impurities - Niobium (Nb) ≤10 ppt
Trace Impurities - Palladium (Pd) ≤20 ppt
Trace Impurities - Platinum (Pt) ≤20 ppt
Trace Impurities - Potassium (K) ≤10 ppt
Trace Impurities - Praseodymium (Pr) ≤1 ppt
Trace Impurities - Rhenium (Re) ≤10 ppt
Trace Impurities - Rhodium (Rh) ≤20 ppt
Trace Impurities - Rubidium (Rb) ≤20 ppt
Trace Impurities - Ruthenium (Ru) ≤20 ppt
Trace Impurities - Samarium (Sm) ≤1 ppt
Trace Impurities - Scandium (Sc) ≤10 ppt
Trace Impurities - Selenium (Se), For Information Only
Trace Impurities - Silver (Ag) ≤10 ppt
Trace Impurities - Sodium (Na) ≤10 ppt
Trace Impurities - Strontium (Sr) ≤10 ppt
Trace Impurities - Tantalum (Ta), For Information Only
Trace Impurities - Tellurium (Te) ≤1 ppt
Trace Impurities - Terbium (Tb) ≤1 ppt
Trace Impurities - Thallium (Tl) ≤10 ppt
Trace Impurities - Thorium (Th) ≤1 ppt
Trace Impurities - Thulium (Tm) ≤1 ppt
Trace Impurities - Tin (Sn) ≤20 ppt
Trace Impurities - Titanium (Ti) ≤20 ppt
Trace Impurities - Tungsten (W) ≤20 ppt
Trace Impurities - Uranium (U) ≤1 ppt
Trace Impurities - Vanadium (V) ≤10 ppt
Trace Impurities - Ytterbium (Yb) ≤1 ppt
Trace Impurities - Yttrium (Y) ≤1 ppt
Trace Impurities - Zinc (Zn) ≤10 ppt
Trace Impurities - Zirconium (Zr) ≤10 ppt
Certificate Provided Reports Actual Lot Analysis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR