Hydrochloric acid 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = NI
BAKR5621-07
BAKR5621-03
5621-02
CAJT5621-7EA
592.85
CAD
CAJT5621-7
CAJT5621-3
JT5621-2
Hydrochloric acid 0.1 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Hydrochloric acid
J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications.
Formula:
HCl Density: 1.004 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,III |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Normality | 0.0995 - 0.1005 |
Sulfate (SO₄) | ≤ 1 ppm |
Ammonium (NH₄) | ≤ 3 ppm |
Trace Impurities - Heavy Metals | ≤ 0.5 ppm |
Trace Impurities - Iron (Fe) | ≤ 0.1 ppm |
Standardization at 25°C traceable to NIST Standard Reference Material. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...