Hydrochloric acid 6 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = N

BAKR5619-02 5619-03
CAJT5619-2CS 915.2 CAD
CAJT5619-2 CAJT5619-3 JT5619-3
Hydrochloric acid 6 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Hydrochloric acid

These acids are tested for common trace metals.

Formula: HCl
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality (eq/L) 5.95 - 6.05
Sulfate (SO₄) ≤ 1 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR