Hydrochloric acid 5 N, BAKER ANALYZED® Volumetric, J.T.Baker®

Supplier: VWR International
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

M.O.C. Category NI

CAJT5618-2 5618-07 5618-03
CAJT5618-2CS 1001.45 CAD
CAJT5618-2 JT5618-7 JT5618-3
Hydrochloric acid 5 N, BAKER ANALYZED® Volumetric, J.T.Baker®
Hydrochloric acid

The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.

Formula: HCl
MW: 36.46
Density: 1.08 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 4.975 - 5.025
Sulfate (SO₄) ≤ 1 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR