Hydrochloric acid 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = N

BAKR5614-03 5614-02
CAJT5614-3CS 851.62 CAD
CAJT5614-3 JT5614-2
Hydrochloric acid 0.02 N, BAKER ANALYZED® volumetric solution, J.T.Baker®
Hydrochloric acid

Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, provided on label.

Formula: HCl
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0195 - 0.0205
Sulfate (SO₄) ≤ 1 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR