Hydrochloric acid 0.01 N, BAKER ANALYZED® Volumetric, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

M.O.C. Category NI

BAKR5611-02 BAKR5611-03 5611-03
CAJT5611-2CS 638.57 CAD
CAJT5611-2 CAJT5611-3 JT5611-3
Hydrochloric acid 0.01 N, BAKER ANALYZED® Volumetric, J.T.Baker®
Hydrochloric acid

Standardized at 25 °C against chemicals whose certification is traceable to NIST. Lot standardization value, accurate to ±1 part per 1000, provided on label.

Formula: HCl
MW: 36.46
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.0095 - 0.0105
Sulfate (SO₄) ≤ 1 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Appearance Passes Test
Standardization at 25°C traceable to NIST Standard Reference Material.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR