1-Hexanesulfonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Sodium hexane-1-sulfonate, Sodium hexane-1-sulphonate, Sodium hexanesulfonate, Sodium hexanesulphonate, Sodium 1-hexanesulfonate, Sodium 1-hexanesulphonate
Management of Change (MOC) category = N
BAKR2175-01
2175-05
CAJT2175-1EA
6182.78
CAD
CAJT2175-1
CAJT2175-5
JT2175-5
1-Hexanesulfonic acid sodium salt ≥98.0% for HPLC, for ion pair chromatography, J.T.Baker®
1-Hexanesulphonic acid sodium salt
Crystals. For ion-pair chromatography of basic compounds.
Formula:
C₆H₁₃O₃SNa MW: 188.22 g/mol Melting Pt: 300 Storage Temperature: Ambient |
MDL Number:
MFCD00007542 CAS Number: 2832-45-3 EINECS: 220-601-3 |
|
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (as C₆H₁₃SO₃Na) (by acidimetry) | ≥98.0 % |
UV Absorbance of 0.25 M Solution - 250 nm | ≤0.050 |
UV Absorbance of 0.25 M Solution - 240 nm | ≤0.050 |
UV Absorbance of 0.25 M Solution - 230 nm | ≤0.050 |
UV Absorbance of 0.25 M Solution - 220 nm | ≤0.060 |
UV Absorbance of 0.25 M Solution - 210 nm | ≤0.080 |
UV Absorbance of 0.25 M Solution - 200 nm | ≤0.200 |
Appearance (clear, colorless, mobile liquid) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...