KIMAX® Erlenmeyer Flasks, Narrow Mouth, Reinforced Beaded Top, Capacity Scale, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
26520-1 26500-4000 26500-6000 26500-125 26500-25 26500-50 26500-250 26500-300 26500-500 26500-1000 26500-2000
89001-342CS 156.67 CAD
89001-342 89087-412 89087-414 89000-782 89001-306 89001-308 89001-310 89001-312 89001-314 89001-316 89001-318
KIMAX® Erlenmeyer Flasks, Narrow Mouth, Reinforced Beaded Top, Capacity Scale, Kimble Chase
Flasks Erlenmeyer Flasks
Versatile flasks for general laboratory use.
  • Tops are reinforced and tooled for maximum mechanical strength
  • Body has thick walls with a long tapered outside contour to minimize chipping
All flasks have durable white ceramic enamel scales to indicate approximate volumes, useful when a high degree of accuracy is not required.
Delivery information: Starter pack contains one flask each of 50, 125, 250, 500, and 1000mL sizes.