Hydrochloric acid 1.0 N, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = R

BAKR0325-07 BAKR0325-09
CAJT0325-7EA 2623.23 CAD
CAJT0325-7 CAJT0325-9
Hydrochloric acid 1.0 N, Biotech Reagent, J.T.Baker®
Hydrochloric acid

Clear, colorless solution. Made from USP purified water and NF (multicompendial) hydrochloric acid that meets BP/Ph. Eur. and JP chemical specifications. Filtered through a 0.2µm filter. GMP manufactured product.


Made from USP purified water and NF (multicompendial) hydrochloric acid that meets BP/Ph. Eur. and JP chemical specifications. GMP manufactured product.


Filtered through a 0.2 micron filter.

Formula: HCl
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 246 °C
Density: 2.71 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Merck Index: 13,08861

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Normality 0.98 - 1.02
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Heavy Metals ≤ 0.5 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Sulfate (SO₄) ≤ 1 ppm
Made from U.S.P. Purified Water and Hydrochloric Acid, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR