Hydrobromic acid 47 - 49% Acidimetry, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hydrogen bromide

M.O.C. Category NI

BAKR0160-03 BAKR0160-01
CAJT0160-3EA 1374.96 CAD
CAJT0160-3 CAJT0160-1
Hydrobromic acid 47 - 49% Acidimetry, BAKER ANALYZED® ACS
Hydrogen bromide

Lot analysis on label.


  • Lot analysis on label

Formula: HBr
MW: 80.91 g/mol
Boiling Pt: -66.7
Melting Pt: -86
Density: 1.50 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8 II
Merck Index: 13,04799

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Material darkens when exposed to air and light.
ACS - Assay (HBr) (by acidimetry) 47.0 - 49.0 %
ACS - Residue after Ignition ≤0.002 %
ACS - Chloride (Cl) ≤0.05 %
ACS - Iodide (I) ≤0.003 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Sulfate and Sulfite (as SO₄) ≤0.003 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤1 ppm
Trace Impurities - Selenium (Se) ≤0.01 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR