Toluene ≥99.8% ACS analytical reagent, general laboratory use

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

LP347-19 LP347-4 LP347-204
CABJLP347D19EA 826.81 CAD
CABJLP347D19 CABJLP347-4 BJLP347-204
Toluene ≥99.8% ACS analytical reagent, general laboratory use
Toluene

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 7.1 %(V)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99.5 % min.
Color (APHA) 10 max.
Residue after Evaporation 0.001 % max.
Substances Darkened by Sulfuric Acid Passes test
Sulfur Compounds (as S) 0.003 % max.
Water by Karl Fischer Titration 0.03 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR