HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid ≥99%, SAFC®
Supplier: MilliporeSigma
1.10110.1000
1.10110.9025
CA1.10110.1000EA
1378.84
CAD
CA1.10110.1000
CA1.10110.9025
HEPES-2-[4-(2-Hydroxyethyl)-1-piperazinyl]-ethanesulfonic acid free acid ≥99%, SAFC®
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)
White crystalline powder. Hygroscopic.
Formula:
C₈H₁₈N₂O₄S Melting Pt: 210…215 °C Density: 1.448 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006158 CAS Number: 7365-45-9 EINECS: 230-907-9 Merck Index: 13,04672 |
|
Specification Test Results
Chloride (Cl) | 0.005% max. |
Heavy Metals (as Pb) | 0.0005% max. |
Loss on Drying (105°C) | 0.2% max. |
Sodium (Na) | 0.005% max. |
Sulfate (SO4) | 0.005% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...