KIMAX® Erlenmeyer Flasks with [ST] Glass Stopper, Graduated, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
26600-25 26600-1000 26600-50 26600-125 26600-250 26600-500
89000-790CS 837.8 CAD
89000-790 89000-792 89001-344 89001-346 89001-348 89001-350
KIMAX® Erlenmeyer Flasks with [ST] Glass Stopper, Graduated, Kimble Chase
Flasks Erlenmeyer Flasks
Flasks are graduated with white scale to indicate approximate volumes and have a [ST] ground glass stopper neck finish.
Designed from ASTM Specification E1404, Type II, Class 2 requirements.