, LabReady™ LC-MS, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

LC407-1 LC407-2.5
CABJLC407-1CS 1398.17 CAD
CABJLC407-1 CABJLC407-2.5
, LabReady™ LC-MS, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Acetonitrile with 0.1% (v/v) trifluoroacetic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.777 g/cm³ (25 °C)
Flash Pt: 6 °C (main component)
Storage Temperature: Ambient
EINECS: 200-929-3
UN: 1648
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR