Hydrochloric acid fuming 37,0 - 38,0%, EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

1.00314.2500 1.00314.9025 1.00314.1000 1.00314.9201 1.00314.1000
CA1.00314.2500EA 316.73 CAD
CA1.00314.2500 CA1.00314.9025 CA1.00314.1000 CA1.00314.9201 EM1.00314.1000
Hydrochloric acid fuming 37,0 - 38,0%, EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP, SAFC®
Hydrochloric acid

Soluble in water. Corrosive and toxic.

Formula: HCl
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Arsenic (As) 0.0001% max.
Boiling Point 110°C (230°F)
Bromide and Iodide (Br, I) Pass Test
Calcium (Ca) 0.001% max.
Free Bromine or Chlorine Pass test
Free Chlorine (Cl) 0.0001% max.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR