Toluene ≥99.8% ACS, meets analytical specification of USP for HPLC, for spectrophotometry

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

AH347-4PC AH347-4
CABJAH347-4PCCS 881.28 CAD
CABJAH347-4PC CABJAH347-4
Toluene ≥99.8% ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Toluene

Meets USP Reagent Specifications


  • ACS certified
  • Lot-to-lot consistency
  • HPLC grade controlled for UV absorbance, GC assay, water content and non-volatile residue

Formula: C₆H₅CH₃
Boiling Pt: 7.1 %(V)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Color (APHA) 10 max.
Water by Karl Fischer Titration 0.03 % max.
GC Analysis 99.8 % min.
Refractive Index (20ºC) 1.4963 - 1.4971
Residue 3 mg/L max.
Substances Darkened by Sulfuric Acid Passes test
Sulfur Compounds (as S) 0.003 % max.
UV Absorbance @ 300 nm 0.1 AU max.
UV Absorbance @ 325 nm 0.025 AU max.
UV Cutoff 284 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR