Hydrofluoric acid 47 - 51%, OmniTrace Ultra™ for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

HX0628-1 HX0628-6
CAHX0628-1EA 985 CAD
CAHX0628-1 CAHX0628-6 CAHX0628-7
Hydrofluoric acid 47 - 51%, OmniTrace Ultra™ for trace metal analysis, Supelco®
Hydrofluoric acid

Clear liquid. For low level trace metal analysis. Double-distilled and packaged in ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom conditions. Supplied in specially designed, preleached fluoropolymer resin bottles to guarantee product integrity.

Formula: HF
MW: 20.01
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790

Order Now

Specification Test Results

Aluminum (Al) 20pg/g max
Antimony (Sb) 20pg/g max
Arsenic (As) 50pg/g max
Assay 47-51%
Barium (Ba) 10pg/g max
Beryllium (Be) 10pg/g max
Bismuth (Bi) 10pg/g max
Boron (B) 100pg/g max
Cadmium (Cd) 10pg/g max
Calcium (Ca) 10pg/g max
Cerium (Ce) 10pg/g max
Cesium (Cs) 10pg/g max
Chromium (Cr) 10pg/g max
Cobalt (Co) 10pg/g max
Copper (Cu) 10pg/g max
Dysprosium (Dy) 1pg/g max
Erbium (Er) 1pg/g max
Europium (Eu) 1pg/g max
Gadolinium (Gd) 1pg/g max
Gallium (Ga) 10pg/g max
Germanium (Ge) 10pg/g max
Gold (Au) 20pg/g max
Hafnium (Hf) 10pg/g max
Holmium (Ho) 1pg/g max
Indium (In) 1pg/g max
Iron (Fe) 10pg/g max
Lanthanum (La) 10pg/g max
Lead (Pb) 10pg/g max
Lithium (Li) 10pg/g max
Lutetium (Lu) 1pg/g max
Magnesium (Mg) 10pg/g max
Manganese (Mn) 10pg/g max
Mercury (Hg) 50pg/g max
Molybdenum (Mo) 10pg/g max
Neodymium (Nd) 1pg/g max
Nickel (Ni) 20pg/g max
Niobium (Nb) 10pg/g max
Palladium (Pd) 20pg/g max
Platinum (Pt) 20pg/g max
Potassium (K) 10pg/g max
Praseodymium (Pr) 1pg/g max
Rhenium (Re) 10pg/g max
Rhodium (Rh) 20pg/g max
Rubidium (Rb) 20pg/g max
Ruthenium (Ru) 20pg/g max
Samarium (Sm) 1pg/g max
Scandium (Sc) 10pg/g max
Silver (Ag) 10pg/g max
Sodium (N/A) 10pg/g max
Strontium (Sr) 10pg/g max
Tellurium (Te) 1pg/g max
Terbium (Tb) 1pg/g max
Thallium (Tl) 10pg/g max
Thorium (Th) 1pg/g max
Thulium (Tm) 1pg/g max
Tin (Sn) 20pg/g max
Titanium (Ti) 20pg/g max
Total sulfur (as S) 100ppb max
Tungsten (W) 20pg/g max
Uranium (U) 1pg/g max
VaN/Adium (V) 10pg/g max
Ytterbium (Yb) 1pg/g max
Yttrium (Y) 1pg/g max
Zinc (Zn) 10pg/g max
Zirconium (Zr) 10pg/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR