Hydrofluoric acid 47 - 51%, OmniTrace® for trace metal analysis, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ100

HX0627-1 HX0627-1
CAHX0627-1CS 12280.28 CAD
EM-HX0627-1 CAHX0627-1
Hydrofluoric acid 47 - 51%, OmniTrace® for trace metal analysis, Supelco®
Hydrofluoric acid

For elemental metal analysis via atomic absorption and ICP instruments. Actual Lot Analysis on label. Color-coded label to match acid cap.

Formula: HF
MW: 20.01 g/mol
MDL Number: MFCD00011346
CAS Number: 7664-39-3
UN: 1790
ADR: 8,II

Order Now

Specification Test Results

Aluminum (Al) 1ppb max
Antimony (Sb) 0.2ppb max
Arsenic (As) 0.5ppb max
Assay 47-51%
Barium (Ba) 0.1ppb max
Beryllium (Be) 0.1ppb max
Bismuth (Bi) 0.1ppb max
Boron (B) 1ppb max
Cadmium (Cd) 0.1ppb max
Calcium (Ca) 1ppb max
Cerium (Ce) 0.1ppb max
Cesium (Cs) 0.1ppb max
Chloride (Cl) 4ppm max
Chromium (Cr) 1ppb max
Cobalt (Co) 0.1ppb max
Color (APHA) 10 max
Copper (Cu) 0.5ppb max
Dysprosium (Dy) 0.1ppb max
Erbium (Er) 0.1ppb max
Europium (Eu) 0.1ppb max
Fluosilicic acid (SiF6) 20ppm max
Gadolinium (Gd) 0.1ppb max
Gallium (Ga) 0.1ppb max
Germanium (Ge) 0.1ppb max
Gold (Au) 0.2ppb max
Hafnium (Hf) 0.1ppb max
Holmium (Ho) 0.1ppb max
Indium (In) 0.1ppb max
Iron (Fe) 1ppb max
Lanthanum (La) 0.1ppb max
Lead (Pb) 0.1ppb max
Lithium (Li) 0.1ppb max
Lutetium (Lu) 0.1ppb max
Magnesium (Mg) 1ppb max
Manganese (Mn) 0.1ppb max
Mercury (Hg) 1ppb max
Molybdenum (Mo) 0.1ppb max
Neodymium (Nd) 0.1ppb max
Nickel (Ni) 0.5ppb max
Niobium (Nb) 0.1ppb max
Palladium (Pd) 0.2ppb max
Platinum (Pt) 0.2ppb max
Potassium (K) 1ppb max
Praseodymium (Pr) 0.1ppb max
Rhenium (Re) 0.1ppb max
Rhodium (Rh) 0.1ppb max
Rubidium (Rb) 0.1ppb max
Ruthenium (Ru) 0.1ppb max
Samarium (Sm) 0.1ppb max
Scandium (Sc) 0.1ppb max
Selenium (Se) 1ppb max
Silver (Ag) 0.5ppb max
Sodium (N/A) 1ppb max
Strontium (Sr) 0.1ppb max
Tellurium (Te) 0.1ppb max
Terbium (Tb) 0.1ppb max
Thallium (Tl) 0.1ppb max
Thorium (Th) 0.1ppb max
Thulium (Tm) 0.1ppb max
Tin (Sn) 0.5ppb max
Titanium (Ti) 1ppb max
Total phosphorus (P) 0.05ppm max
Total sulfur (as S) 0.1ppm max
Tungsten (W) 0.5ppb max
Uranium (U) 0.1ppb max
VaN/Adium (V) 0.1ppb max
Ytterbium (Yb) 0.1ppb max
Yttrium (Y) 0.1ppb max
Zinc (Zn) 1ppb max
Zirconium (Zr) 0.1ppb max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR