PYREX® Wide Mouth Media Storage Bottles, Corning
Supplier: Corning
1397-1L 1397-2L 1397-500
89090-054CS 489.52 CAD
89090-054 89090-060 89090-062
PYREX® Wide Mouth Media Storage Bottles, Corning
Bottles Cell Culture Bottles
Made of PYREX® borosilicate glass.
  • Resistant to thermal shock
  • Permanent white enamel graduations and marking spot
  • Traceability Code for easier documentation
  • Not recommended for vaccum use
Wide mouth media storage bottles are designed for heavy duty storage as well as mixing and sampling. They includes linerless, one-piece autoclavable orange polypropylene plug seal GLS80-threaded cap with a drip-free pouring ring.
Caution: Do not tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. These bottles shouldMore Product Information