Toluene ≥99.8%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Methylbenzene, Phenylmethane, FM approval
347-1L
347-4
347-4
CABJ347-1LCS
526.27
CAD
CABJ347-1L
CABJ347-4
CABJNP347-20
BJ347-4
Toluene ≥99.8%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Toluene
For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.
- Minimal UV contaminants
- High GC purity
- Low water content
- Outstanding lot-to-lot consistency
Formula:
C₆H₅CH₃ MW: 92.14 g/mol Boiling Pt: 110.6 °C Melting Pt: –95 °C Density: 0.867 g/cm³ (20 °C) Flash Pt: 7.1 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3,II REACH: 05-2114615130-69 Merck Index: 13,09607 |
Specification Test Results
Benzene | 500 mg/L max. |
Electron Capture GC | 2 µg/L max. |
GC Analysis | 99.8 % min. |
Refractive Index (20ºC) | 1.4963 - 1.4971 |
Residue | 1 mg/L max. |
UV Absorbance @ 300 nm | 0.12 AU max. |
UV Absorbance @ 325 nm | 0.025 AU max. |
UV Cutoff | 284 nm max. |
Water by Karl Fischer Titration | 0.03 % max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...