Tetrahydrofuran 99,9 GC_ASSAY_METHOD Stabilised, B&J Brand™ ACS GPC_GC, Spectrophotometry_non, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

Stabilization: 250 ppm BHT
Nitrogen_filled

341-1L 341-4
CABJ341-1LCS 1111.21 CAD
CABJ341-1L CABJ341-4
Tetrahydrofuran 99,9 GC_ASSAY_METHOD Stabilised, B&J Brand™ ACS GPC_GC, Spectrophotometry_non, Burdick & Jackson™
Tetrahydrofuran

For GC and GPC. Non-UV grade. Packed under nitrogen. Preserved with 250ppm BHT.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency


200-liter Stainless Steel Pressure Dispense System (SSPDS) containers that are returned for refill. Sold only under contract.

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.8892 g/cm³ (20 °C)
Flash Pt: 11.8 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.9 % min.
Peroxides (as H₂O₂) at time of packaging 2 mg/L max.
Refractive Index (20ºC) 1.4065 - 1.4075
Water by Karl Fischer Titration 0.03 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR