Tetrahydrofuran ≥99.9% contains no preservatives, B&J Brand™ ACS, gradient grade for HPLC, for spectrophotometry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

NS340-20 340-1L 340-4 NS340-20 CS340-200
CABJNS340-20EA 2582.83 CAD
CABJNS340-20 CABJ340-1L CABJ340-4 BJNS340-20 BJCS340-200
Tetrahydrofuran ≥99.9% contains no preservatives, B&J Brand™ ACS, gradient grade for HPLC, for spectrophotometry, Burdick & Jackson™
Tetrahydrofuran

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C
Melting Pt: –108.5 °C
Density: 0.8892 g/cm³ (20 °C)
Flash Pt: 11.8 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.9 % min.
Water by Karl Fischer Titration 0.03 % max.
Peroxides (as H₂O₂) at time of packaging 2 mg/L max.
Refractive Index (20ºC) 1.4065 - 1.4075
Residue 1 mg/L max.
UV Absorbance @ 250 nm 0.18 AU max.
UV Absorbance @ 300 nm 0.02 AU max.
UV Absorbance @ 350 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 212 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR