Synthware Recovery Flasks, Heavy-Wall, Kemtech America
Supplier: Kemtech America
F311405 F311410 F311910 F311425 F311925 F312425 F311450 F311950 F312450 F311100 F319100 F314100 F318100 F311200 F314200 F311250 F319250 F314250 F318250 F314500 F318500 F314000 F31291L
60002-410EA 45.93 CAD
60002-410 60002-412 60002-414 60002-416 60002-418 60002-420 60002-422 60002-424 60002-426 60002-428 60002-430 60002-432 60002-434 60002-436 60002-440 60002-444 60002-446 60002-448 60002-450 60002-452 60002-454 60002-456 60002-458
Synthware Recovery Flasks, Heavy-Wall, Kemtech America
Flasks Distillation Flasks
Hand-blown from tubing to ensure uniform wall thickness.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Various sizes available
The Synthware recovery flask is designed for heating. Evaporation and storage or transfer of solvents.