Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
HX0603-23
HX0603-3
HX0603-4
HX0603-75
HX0603P-5
CAHX0603-23EA
1804
CAD
CAHX0603-23
CAHX0603-3
CAHX0603-4
CAHX0603-75
CAHX0603P-5
Hydrochloric acid 36.5 - 38.0%, GR ACS, Supelco®
Formula:
HCl Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II |
Specification Test Results
Aluminum (Al) | 0.3ppm max |
Ammonium (NH4) | 3ppm max |
Appearance | To pass test |
Arsenic (As) | 0.01ppm max |
Assay | 36.5-38.0% |
Bromide (Br) | 0.005% max |
Calcium (Ca) | 0.3ppm max |
Chromium (Cr) | 0.2ppm max |
Color (APHA) | 10 max |
Copper (Cu) | 0.1ppm max |
Free chlorine (as Cl) | 1ppm max |
Gold (Au) | 0.3ppm max |
Heavy metals (as Pb) | 1ppm max |
Iron (Fe) | 0.2ppm max |
Lead (Pb) | 0.1ppm max |
Magnesium (Mg) | 0.3ppm max |
Manganese (Mn) | 0.3ppm max |
Mercury (Hg) | 0.005ppm max |
Nickel (Ni) | 0.1ppm max |
Potassium (K) | 0.3ppm max |
Residue after ignition | 5ppm max |
Sodium (N/A) | 0.3ppm max |
Sulfate (SO4) | 1ppm max |
Sulfite (SO3) | 1ppm max |
Tin (Sn) | 0.3ppm max |
Titanium (Ti) | 0.3ppm max |
Zinc (Zn) | 0.3ppm max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...