Hexane (mixture of isomers) ≥98%, Practical grade, Supelco®

Supplier: MilliporeSigma
Danger

HX0300-3 HX0300-1
CAHX0300-3EA 1240 CAD
CAHX0300-3 CAHX0300-1
Hexane (mixture of isomers) ≥98%, Practical grade, Supelco®
Hexane (mixture of isomers)

Practical

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (n-hexane) Report
Assay (total hexane isomers + methylcyclopentane) 98% min
Color (APHA) 20 max
Form Clear liquid
Infrared spectrum Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR