Hexane (mixture of isomers) ≥98.5% (total hexane isomers + methylcyclopentane) n-Hexane ∼ 64% ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Hexanes, Isohexane
HX0299-20
HX0299-3
HX0299-20
HX0299-5
HX0299-6
HX0299P-4
CAHX0299-20EA
3305
CAD
CAHX0299-20
CAHX0299-3
EM-HX0299-20
CAHX0299-5
CAHX0299-6
CAHX0299P-4
CA-HX0299T-4
Hexane (mixture of isomers) ≥98.5% (total hexane isomers + methylcyclopentane) n-Hexane ∼ 64% ACS, Supelco®
Hexane (mixture of isomers)
Meets ACS Specifications
Formula:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Boiling Pt: 69 °C (1013 hPa) Melting Pt: -95 Density: 0.66 g/cm3 (20 °C) Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179311 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3 II Merck Index: 13,04712 |
|
Specification Test Results
Assay (n-hexane) | Report |
Assay (total hexane isomers + methylcyclopentane) | 98.5% min |
Color (APHA) | 10 max |
Form | Clear liquid |
Residue after evaporation | 0.001% max |
Sulfur compounds (as S) | 0.005% max |
Thiophene | To pass test |
Water-soluble titrable acid | 0.0003meq/g max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...