Hexane (mixture of isomers) ≥98.5%, OmniSolv®, non-UV for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexanes, Isohexane

HX0298-1 HX0298-6
CAHX0298-1CS 1604 CAD
CAHX0298-1 CAHX0298-6
Hexane (mixture of isomers) ≥98.5%, OmniSolv®, non-UV for gas chromatography, Supelco®
Hexane (mixture of isomers)

For gas chromatography and residue analysis. Not optically transparent in the UV region. Purity based on n-hexane and saturated C6 hydrocarbons. Lot analysis on label.

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (n-hexane) Report
Assay (total aliphatic hydrocarbons C8) 99.8% min
Assay (total hexane isomers + methylcyclopentane) 98.5% min
Boiling range 66.1-69.4°C
Capillary ECD responsive substances (as PCNB) 2ng/L max
Capillary FID responsive substances (as decane) 3µg/L max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Infrared spectrum Conforms
Residue after evaporation 1ppm max
Water (H2O) 0.01% max

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR