n-Hexane ≥95%, OmniSolv®, Supelco®
Supplier: MilliporeSigma
|
Danger
|
HX0295P-1
HX0295-1
HX0295-6
CAHX0295P-1EA
628
CAD
CAHX0295P-1
CAHX0295-1
CAHX0295-6
n-Hexane ≥95%, OmniSolv®, Supelco®
n-Hexane
Suitable for spectrophotometry, HPLC, and gas chromatography and residue analysis. UV cutoff 195nm. Certificate of Analysis on label.
Formula:
H₃C(CH₂)₄CH₃ Boiling Pt: 69 °C (1013 hPa) Melting Pt: –94.3 °C Density: 0.659 g/cm³ (20 °C) Flash Pt: –22 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009520 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 13,04712 |
|
Specification Test Results
Assay (n-hexane) | 95% min |
Boiling range | 66.1-69.4°C |
Capillary ECD responsive substances (as PCNB) | 5ng/L max |
Capillary FID responsive substances (as decane) | 3µg/L max |
Color (APHA) | 10 max |
Filtered through 0.2 µm filter | To pass test |
Fluorescence (as quinine base) | 250ppt max |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Residue after evaporation | 1ppm max |
UV Abs. at 195 nm | 1.00AU max |
UV Abs. at 200 nm | 0.50AU max |
UV Abs. at 210 nm | 0.20AU max |
UV Abs. at 220 nm | 0.05AU max |
UV Abs. at 230 nm | 0.05AU max |
UV Abs. at 250 nm | 0.01AU max |
UV Abs. at 280 nm | 0.005AU max |
UV Cut-off | 195nm max |
Water (H2O) | 0.01% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...